Thermo Scientific Chemicals

2-(4-Nitrophenyl)ethylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: H64894.03
1 g, Each
Thermo Scientific Chemicals

2-(4-Nitrophenyl)ethylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: H64894.03
1 g, Each
Quantity
Catalog number: H64894.03
also known as H64894-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29968-78-3
IUPAC Name
2-(4-nitrophenyl)ethan-1-aminium
Molecular Formula
C8H11N2O2
InChI Key
IOXOZOPLBFXYLM-UHFFFAOYSA-O
SMILES
[NH3+]CCC1=CC=C(C=C1)[N+]([O-])=O
Assay (HPLC)
>98.0%

Description

4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.

Solubility
Soluble in methanol almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text