5-Bromo-2-chloro-3-nitropyridine, 98%, Thermo Scientific Chemicals
5-Bromo-2-chloro-3-nitropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-chloro-3-nitropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H64897.14
also known as H64897-14
Price (USD)/ Each
67.70
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
67.70
Add to cart
5-Bromo-2-chloro-3-nitropyridine, 98%, Thermo Scientific Chemicals
Catalog numberH64897.14
Price (USD)/ Each
67.70
-
Add to cart
Chemical Identifiers
CAS67443-38-3
IUPAC Name5-bromo-2-chloro-3-nitropyridine
Molecular FormulaC5H2BrClN2O2
InChI KeyWWQQPSDIIVXFOX-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Br)=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only
A. H. Berrie.; G. T. Newbold.; F. S. Spring. Some reactions of substituted 2-bromopyridines.J. Chem. Soc. 1952, 2042-2046 .