Thermo Scientific Chemicals

Dibenzofuran-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H64905.03
1 g, Each
Thermo Scientific Chemicals

Dibenzofuran-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H64905.03
1 g, Each
Quantity
Catalog number: H64905.03
also known as H64905-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5397-82-0
IUPAC Name
8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-4-carbaldehyde
Molecular Formula
C13H8O2
InChI Key
OVJMIWIVPWPZMN-UHFFFAOYSA-N
SMILES
O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1
Appearance (Color)
Pale cream to pale yellow
Form
Powder
Assay from Supplier's CofA
≥97.5%

Description

Used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an pharmaceutical intermediate.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text