3-Fluoro-2-formylbenzeneboronic acid, 95%, Thermo Scientific Chemicals
3-Fluoro-2-formylbenzeneboronic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-2-formylbenzeneboronic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number H64909.14
also known as H64909-14
Price (USD)/ Each
776.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
776.00
Add to cart
3-Fluoro-2-formylbenzeneboronic acid, 95%, Thermo Scientific Chemicals
Catalog numberH64909.14
Price (USD)/ Each
776.00
-
Add to cart
Chemical Identifiers
CAS871126-15-7
IUPAC Name(3-fluoro-2-formylphenyl)boronic acid
Molecular FormulaC7H6BFO3
InChI KeyGVHWLCYABLWGIR-UHFFFAOYSA-N
SMILESOB(O)C1=C(C=O)C(F)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
Undergoes unprecedented tautomeric rearrangement in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undergoes unprecedented tautomeric rearrangement in solution.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Sergiusz Luliński; Janusz Serwatowski. A diverse structural behaviour of boronated ortho-phthalaldehydes: A crystal structure of 1,3-dihydro-1,3-dihydroxy-4-formylbenzo[c][2,1]oxaborole. Journal of Organometallic Chemistry.2007 692(14), 2924-2929.
  2. Jiyoung Yang; Brandon J. Johnson; Allan A. Letourneau; Christopher M. Vogels; Andreas Decken; Felix J. Baerlocher; and Stephen A. Westcott.Synthesis, Characterisation, and Antifungal Activities of Novel Benzodiazaborines. Australian Journal of Chemistry.2014 68(3), 366-372.