D-Proline ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
D-Proline ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Proline ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H64910.14
also known as H64910-14
Price (USD)/ Each
220.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
220.00
Add to cart
D-Proline ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH64910.14
Price (USD)/ Each
220.00
-
Add to cart
Chemical Identifiers
CAS131477-20-8
IUPAC Nameethyl pyrrolidine-2-carboxylate hydrochloride
Molecular FormulaC7H14ClNO2
InChI KeyDUJGQVVONTYHLT-UHFFFAOYNA-N
SMILESCl.CCOC(=O)C1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Assay (HPLC)>97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only