10-Phenylanthracene-9-boronic acid, 98%, Thermo Scientific Chemicals
10-Phenylanthracene-9-boronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

10-Phenylanthracene-9-boronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64911.03
also known as H64911-03
Price (USD)/ Each
93.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
93.80
Add to cart
10-Phenylanthracene-9-boronic acid, 98%, Thermo Scientific Chemicals
Catalog numberH64911.03
Price (USD)/ Each
93.80
-
Add to cart
Chemical Identifiers
CAS334658-75-2
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is used as pharmaceutical intermediate and as OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as OLED materials.

Solubility
Soluble in methanol.

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only