Thermo Scientific Chemicals

10-Phenylanthracene-9-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H64911.06
5 g, Each
Thermo Scientific Chemicals

10-Phenylanthracene-9-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H64911.06
5 g, Each
Quantity
Catalog number: H64911.06
also known as H64911-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
334658-75-2
Assay (HPLC)
>97.5%

Description

It is used as pharmaceutical intermediate and as OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as OLED materials.

Solubility
Soluble in methanol.

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text