Thermo Scientific Chemicals

2-(4-Bromophenyl)-2-propanol, 97%, Thermo Scientific Chemicals

Catalog number: H64919.03
1 g, Each
Thermo Scientific Chemicals

2-(4-Bromophenyl)-2-propanol, 97%, Thermo Scientific Chemicals

Catalog number: H64919.03
1 g, Each
Quantity
Catalog number: H64919.03
also known as H64919-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2077-19-2
IUPAC Name
2-(4-bromophenyl)propan-2-ol
Molecular Formula
C9H11BrO
InChI Key
AOGYBHJTXLXRSM-UHFFFAOYSA-N
SMILES
CC(C)(O)C1=CC=C(Br)C=C1
Assay (HPLC)
≥96.0%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.

Solubility
Slightly soluble in water (1.4 g/L) (25°C)

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text