N-Boc-1,4-diaminocyclohexane, 95%, Thermo Scientific Chemicals
N-Boc-1,4-diaminocyclohexane, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-1,4-diaminocyclohexane, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H64932.06
also known as H64932-06
Price (USD)/ Each
471.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
471.00
Add to cart
N-Boc-1,4-diaminocyclohexane, 95%, Thermo Scientific Chemicals
Catalog numberH64932.06
Price (USD)/ Each
471.00
-
Add to cart
Chemical Identifiers
CAS195314-59-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only