Dimethyl pyridine-2,3-dicarboxylate, 95%, Thermo Scientific Chemicals
Dimethyl pyridine-2,3-dicarboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl pyridine-2,3-dicarboxylate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H64939.14
also known as H64939-14
Price (USD)/ Each
91.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
91.90
Add to cart
Dimethyl pyridine-2,3-dicarboxylate, 95%, Thermo Scientific Chemicals
Catalog numberH64939.14
Price (USD)/ Each
91.90
-
Add to cart
Chemical Identifiers
CAS605-38-9
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only