Thermo Scientific Chemicals

3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64945.06
5 g, Each
Thermo Scientific Chemicals

3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64945.06
5 g, Each
Quantity
Catalog number: H64945.06
also known as H64945-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
72587-18-9
IUPAC Name
2-chloro-5-(trifluoromethyl)pyridin-3-amine
Molecular Formula
C6H4ClF3N2
InChI Key
HABKYPVMMHBOIW-UHFFFAOYSA-N
SMILES
NC1=C(Cl)N=CC(=C1)C(F)(F)F
Assay (HPLC)
>97.5%

Description

It's employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It’s employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text