Thermo Scientific Chemicals

Copper(I) thiophene-2-carboxylate, Thermo Scientific Chemicals

Catalog number: H64950.06
5 g, Each
Thermo Scientific Chemicals

Copper(I) thiophene-2-carboxylate, Thermo Scientific Chemicals

Catalog number: H64950.06
5 g, Each
Quantity
Catalog number: H64950.06
also known as H64950-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68986-76-5
IUPAC Name
λ¹-copper(1+) thiophene-2-carboxylate
Molecular Formula
C5H3CuO2S
InChI Key
SFJMFSWCBVEHBA-UHFFFAOYSA-M
SMILES
[Cu+].[O-]C(=O)C1=CC=CS1
Assay (special)
≥80.0% (Carbon EA, non-U.S. specification)
Comment
Purchased in the U.S. and in other countries
Elemental Analysis
Carbon, C: 25.19-37.79% (Theory 31.49%) (non-U.S. specification)
Elemental Analysis
Copper, Cu: 26.66-39.99% (Theory 33.32%) (non-U.S. specification)
Appearance (Color)
Yellow to green or red to brown

Description

Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.

Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text