Thermo Scientific Chemicals

2-Bromo-5-methoxybenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: H64963.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-5-methoxybenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: H64963.06
5 g, Each
Quantity
Catalog number: H64963.06
also known as H64963-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19614-12-1
IUPAC Name
1-bromo-2-(bromomethyl)-4-methoxybenzene
Molecular Formula
C8H8Br2O
InChI Key
MURVUTUZSUEIGI-UHFFFAOYSA-N
SMILES
COC1=CC(CBr)=C(Br)C=C1
Assay (HPLC)
>96.0%

Description

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Soluble in hot methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text