Thermo Scientific Chemicals

2-Bromo-3'-chloropropiophenone, 98%, Thermo Scientific Chemicals

Catalog number: H64999.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-3'-chloropropiophenone, 98%, Thermo Scientific Chemicals

Catalog number: H64999.06
5 g, Each
Quantity
Catalog number: H64999.06
also known as H64999-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34911-51-8
IUPAC Name
2-bromo-1-(3-chlorophenyl)propan-1-one
Molecular Formula
C9H8BrClO
InChI Key
OFNMQTRHMBQQEA-UHFFFAOYNA-N
SMILES
CC(Br)C(=O)C1=CC(Cl)=CC=C1
Assay (HPLC)
>97.5%

Description

It used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in acetonitrile, chloroform, dichloromethane and ethyl acetate.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text