N-Benzyloxycarbonyl-L-glutaminylglycine, 98%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-glutaminylglycine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-glutaminylglycine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number H65088.03
also known as H65088-03
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
Add to cart
N-Benzyloxycarbonyl-L-glutaminylglycine, 98%, Thermo Scientific Chemicals
Catalog numberH65088.03
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6610-42-0
IUPAC Name2-(2-{[(benzyloxy)carbonyl]amino}-4-carbamoylbutanamido)acetic acid
Molecular FormulaC15H19N3O6
InChI KeySOUXAAOTONMPRY-UHFFFAOYNA-N
SMILESNC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream
Assay from Suppliers CofA≥97.5% (HPLC)
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only