D-Histidine methyl ester dihydrochloride, 95%, Thermo Scientific Chemicals
D-Histidine methyl ester dihydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Histidine methyl ester dihydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H65096.06
also known as H65096-06
Price (USD)/ Each
101.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
101.00
Add to cart
D-Histidine methyl ester dihydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH65096.06
Price (USD)/ Each
101.00
-
Add to cart
Chemical Identifiers
CAS4467-54-3
IUPAC Namemethyl 2-amino-3-(1H-imidazol-5-yl)propanoate dihydrochloride
Molecular FormulaC7H13Cl2N3O2
InChI KeyDWAYENIPKPKKMV-UHFFFAOYNA-N
SMILESCl.Cl.COC(=O)C(N)CC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only