4-Chloropyridine-2-carboxamide, 97%, Thermo Scientific Chemicals
4-Chloropyridine-2-carboxamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloropyridine-2-carboxamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Catalog number H65265.22
also known as H65265-22
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS99586-65-9
IUPAC Name4-chloropyridine-2-carboxamide
Molecular FormulaC6H5ClN2O
InChI KeyXIHHOUUTBZSYJH-UHFFFAOYSA-N
SMILESNC(=O)C1=CC(Cl)=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only