Thermo Scientific Chemicals

beta-Alaninamide hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H65365.06
5 g, Each
Thermo Scientific Chemicals

beta-Alaninamide hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H65365.06
5 g, Each
Quantity
Catalog number: H65365.06
also known as H65365-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64017-81-8
IUPAC Name
3-aminopropanamide hydrochloride
Molecular Formula
C3H9ClN2O
InChI Key
GAGJMOQGABUOBK-UHFFFAOYSA-N
SMILES
Cl.NCCC(N)=O
Assay (HPLC)
>95.0%

Description

beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat , air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text