Thermo Scientific Chemicals

3-Dimethylaminopropyl chloride, 97%, stab. with 0.5% oxalic acid, Thermo Scientific Chemicals

Catalog number: H65371.03
1 g, Each
Thermo Scientific Chemicals

3-Dimethylaminopropyl chloride, 97%, stab. with 0.5% oxalic acid, Thermo Scientific Chemicals

Catalog number: H65371.03
1 g, Each
Quantity
Catalog number: H65371.03
also known as H65371-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-54-6
IUPAC Name
hydrogen (3-chloropropyl)dimethylamine chloride
Molecular Formula
C5H13Cl2N
InChI Key
LJQNMDZRCXJETK-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CCCCl
Identification (FTIR)
Conforms
Proton NMR
Conforms
Assay (GC)
≥96%
Refractive Index
1.43
Form
Liquid

Description

It is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text