N-Glycyl-L-tyrosine, 98%, Thermo Scientific Chemicals
N-Glycyl-L-tyrosine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Glycyl-L-tyrosine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number H65388.06
also known as H65388-06
Price (USD)/ Each
70.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
70.70
Add to cart
N-Glycyl-L-tyrosine, 98%, Thermo Scientific Chemicals
Catalog numberH65388.06
Price (USD)/ Each
70.70
-
Add to cart
Chemical Identifiers
CAS658-79-7
IUPAC Name(2S)-2-(2-aminoacetamido)-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC11H14N2O4
InChI KeyXBGGUPMXALFZOT-IMWMWJONNA-N
SMILESNCC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.9%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only