D-Leucine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
D-Leucine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Leucine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number H65456.22
also known as H65456-22
Price (USD)/ Each
990.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
990.00
Add to cart
D-Leucine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH65456.22
Price (USD)/ Each
990.00
-
Add to cart
Chemical Identifiers
CAS5845-53-4
IUPAC Namemethyl (2R)-2-amino-4-methylpentanoate hydrochloride
Molecular FormulaC7H16ClNO2
InChI KeyDODCBMODXGJOKD-HQFIOVGONA-N
SMILESCl.COC(=O)[C@H](N)CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
Appearance (Color)White to off-white
Assay from Supplier's CofA≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only