4-Dodecylaniline has been used: in the synthesis of sodium 4-dodecylphenylazosulfonate, to investigate the water solubilization of single-walled carbon nanotubes (SWNTs), in proton transfer step during molecular oxygen (O2) reduction by decamethylferrocene (DMFc).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Dodecylaniline has been used: in the synthesis of sodium 4-dodecylphenylazosulfonate, to investigate the water solubilization of single-walled carbon nanotubes (SWNTs), in proton transfer step during molecular oxygen (O2) reduction by decamethylferrocene (DMFc).
Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Ian R. Dunkin; Andreas Gittinger; David C. Sherrington and Paul Whittaker. A photodestructible surfactant. J. Chem. Soc., Chem. Commun.1994, 2245-2246.
Francisco Pompeo and Daniel E. Resasco. Water Solubilization of Single-Walled Carbon Nanotubes by Functionalization with Glucosamine. Nano Letters.2002, 2, (4), 369-373.