N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester, 95%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H65546.06
also known as H65546-06
Price (USD)/ Each
49.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
49.80
Add to cart
N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester, 95%, Thermo Scientific Chemicals
Catalog numberH65546.06
Price (USD)/ Each
49.80
-
Add to cart
Chemical Identifiers
CAS3397-35-1
IUPAC Name2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoate
Molecular FormulaC18H22N2O6
InChI KeyYHZUOMRURVTBMO-YQTOOIBONA-N
SMILESCC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)≥94.0%
Melting Point116-120?C
Optical Rotation|0 = -33? ?2? (C=1 in Dioxane)
Solution Test2 mmol in 1 ml DCM + 1 ml DMF
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only