A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Horiike K; Nishina Y; Miyake Y; Yamano T. Affinity labeling of D-amino acid oxidase with an acetylenic substrate. J Biochem.1975 78(1), 57-63.
Patrick Marcotte; Christopher Walsh.Properties of D-amino acid oxidase covalently modified upon its oxidation of D-propargylglycine. Biochemistry.1978 17(14), 2864-2868.