N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester, 95%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-(N-succinimidyl) ester, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H65816.06
also known as H65816-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3338-32-7
IUPAC Name1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3-{[(benzyloxy)carbonyl]amino}butanedioate
Molecular FormulaC20H24N2O8
InChI KeyJUMSBOKRGDETHL-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only