Thermo Scientific Chemicals

N-Isopropylmethylamine, 98%, Thermo Scientific Chemicals

Catalog number: H65892.14
25 g, Each
Thermo Scientific Chemicals

N-Isopropylmethylamine, 98%, Thermo Scientific Chemicals

Catalog number: H65892.14
25 g, Each
Quantity
Catalog number: H65892.14
also known as H65892-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4747-21-1
IUPAC Name
methyl(propan-2-yl)amine
Molecular Formula
C4H11N
InChI Key
XHFGWHUWQXTGAT-UHFFFAOYSA-N
SMILES
CNC(C)C
Assay (HPLC)
>97.5%

Description

N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.

Solubility
Soluble in chloroform, DMSO, methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Do not store together with acids. Incompatible with acids , air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text