D-Glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals
D-Glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H65903.14
also known as H65903-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS79338-14-0
IUPAC Name4-amino-5-(benzyloxy)-5-oxopentanoic acid
Molecular FormulaC12H15NO4
InChI KeyHFZKKJHBHCZXTQ-UHFFFAOYNA-N
SMILESNC(CCC(O)=O)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only