tert-Butyl 4-aminobutyrate hydrochloride, 97%, Thermo Scientific Chemicals
tert-Butyl 4-aminobutyrate hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl 4-aminobutyrate hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H65936.03
also known as H65936-03
Price (USD)/ Each
60.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
60.00
Add to cart
tert-Butyl 4-aminobutyrate hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH65936.03
Price (USD)/ Each
60.00
-
Add to cart
Chemical Identifiers
CAS58640-01-0
IUPAC Nametert-butyl 4-aminobutanoate hydrochloride
Molecular FormulaC8H18ClNO2
InChI KeyCZVNLCOJFFYZPG-UHFFFAOYSA-N
SMILESCl.CC(C)(C)OC(=O)CCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
Employed as an important intermediate for raw material for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for pharmaceutical.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
G Di Silvestro; G Palmisano; R Pellegata. Phase diagram of (R)- and (S)-4-hydroxy-2-pyrrolidone mixtures: a new case of a conglomerate-forming system. Journal of Pharmaceutical Sciences.1993 82(7), 758-760.