2-Acetyl-6-methylpyridine, 98%, Thermo Scientific Chemicals
2-Acetyl-6-methylpyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Acetyl-6-methylpyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H65997.03
also known as H65997-03
Price (USD)/ Each
89.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
89.50
Add to cart
2-Acetyl-6-methylpyridine, 98%, Thermo Scientific Chemicals
Catalog numberH65997.03
Price (USD)/ Each
89.50
-
Add to cart
Chemical Identifiers
CAS6940-57-4
IUPAC Name1-(6-methylpyridin-2-yl)ethan-1-one
Molecular FormulaC8H9NO
InChI KeyFPQMUQPPAYCAME-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(C)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only