Specifications
Chemical Name or Material(3-Bromophenylethynyl)trimethylsilane
CAS3989-13-7
Health Hazard 1H315-H319-H335
Health Hazard 2H315-H319-H335; Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more