Thermo Scientific Chemicals

(S)-(+)-3-Aminotetrahydrofuran hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H66034.MD
250 mg, Each
Thermo Scientific Chemicals

(S)-(+)-3-Aminotetrahydrofuran hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H66034.MD
250 mg, Each
Quantity
Catalog number: H66034.MD
also known as H66034-MD
Price (USD)
Have Questions?

Chemical Identifiers

CAS
204512-95-8
IUPAC Name
oxolan-3-amine hydrochloride
Molecular Formula
C4H10ClNO
InChI Key
MHOVLDXJDIEEMJ-UHFFFAOYNA-N
SMILES
Cl.NC1CCOC1
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥94.0%
Form
Crystalline powder or powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text