6-Fluoroquinoline-2-carboxaldehyde, 97%, Thermo Scientific Chemicals
6-Fluoroquinoline-2-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Fluoroquinoline-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H66052.MD
also known as H66052-MD
Price (USD)/ Each
46.70
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
46.70
Add to cart
6-Fluoroquinoline-2-carboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH66052.MD
Price (USD)/ Each
46.70
-
Add to cart
Chemical Identifiers
CAS260430-93-1
IUPAC Name6-fluoroquinoline-2-carbaldehyde
Molecular FormulaC10H6FNO
InChI KeyJDQMJKKGPXPPOJ-UHFFFAOYSA-N
SMILESFC1=CC=C2N=C(C=O)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow
Assay (GC)>96.0%
FormPowder
Free acid (titration)<1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only