Thermo Scientific Chemicals

2-Bromo-3'-chloroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: H66077.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-3'-chloroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: H66077.06
5 g, Each
Quantity
Catalog number: H66077.06
also known as H66077-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
41011-01-2
IUPAC Name
2-bromo-1-(3-chlorophenyl)ethan-1-one
Molecular Formula
C8H6BrClO
InChI Key
KJVRURZDIOVSSQ-UHFFFAOYSA-N
SMILES
ClC1=CC=CC(=C1)C(=O)CBr
Assay (HPLC)
>96.0%

Description

2-Bromo-3'-chloroacetophenone acts as a Substrate used in a one-step preparation of symmetrical 1,4-diketones in the presence of Zn-I2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3′-chloroacetophenone acts as a Substrate used in a one-step preparation of symmetrical 1,4-diketones in the presence of Zn-I2.

Notes
Store at 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text