Thermo Scientific Chemicals

4-Bromoresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: H66087.06
5 g, Each
Thermo Scientific Chemicals

4-Bromoresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: H66087.06
5 g, Each
Quantity
Catalog number: H66087.06
also known as H66087-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6626-15-9
IUPAC Name
4-bromobenzene-1,3-diol
Molecular Formula
C6H5BrO2
InChI Key
MPCCNXGZCOXPMG-UHFFFAOYSA-N
SMILES
OC1=CC=C(Br)C(O)=C1
Form
Powder or crystals or crystalline powder
Appearance (Color)
Pale brown to pale pink
Assay (HPLC)
≥97.5% (GC)

Description

It is commonly used as an organic reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is commonly used as an organic reagent and pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text