1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%, Thermo Scientific Chemicals
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H66108.03
also known as H66108-03
Price (USD)/ Each
405.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
405.00
Add to cart
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%, Thermo Scientific Chemicals
Catalog numberH66108.03
Price (USD)/ Each
405.00
-
Add to cart
Chemical Identifiers
CAS3849-01-2
IUPAC Name1,3,5-tris(chloromethyl)-2,4,6-trimethylbenzene
Molecular FormulaC12H15Cl3
InChI KeyPHQFMPNZCIHSPC-UHFFFAOYSA-N
SMILESCC1=C(CCl)C(C)=C(CCl)C(C)=C1CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only