Thermo Scientific Chemicals

2-Amino-4-methoxypyridine, 95%, Thermo Scientific Chemicals

Catalog number: H66125.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-4-methoxypyridine, 95%, Thermo Scientific Chemicals

Catalog number: H66125.14
25 g, Each
Quantity
Catalog number: H66125.14
also known as H66125-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10201-73-7
IUPAC Name
4-methoxypyridin-2-amine
Molecular Formula
C6H8N2O
InChI Key
QPHBCOSULYSASF-UHFFFAOYSA-N
SMILES
COC1=CC(N)=NC=C1
Assay (HPLC)
>94.0%

Description

2-Amino-4-methoxypyridine is a highly selective inducible nitric oxide synthase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methoxypyridine is a highly selective inducible nitric oxide synthase inhibitors.

Solubility
Soluble in Dimethyl sulfoxide, methanol. Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text