8-Bromo-2-chloro-6-fluoroquinazoline, 95%, Thermo Scientific Chemicals
8-Bromo-2-chloro-6-fluoroquinazoline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Bromo-2-chloro-6-fluoroquinazoline, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H66128.03
also known as H66128-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS953039-63-9
IUPAC Name8-bromo-2-chloro-6-fluoroquinazoline
Molecular FormulaC8H3BrClFN2
InChI KeyXTEWVYPXKPYPRE-UHFFFAOYSA-N
SMILESFC1=CC(Br)=C2N=C(Cl)N=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)99.40%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only