2-Bromo-5-cyanopyridine, 97%, Thermo Scientific Chemicals
2-Bromo-5-cyanopyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-5-cyanopyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66177.03
also known as H66177-03
Price (USD)/ Each
39.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
39.20
Add to cart
2-Bromo-5-cyanopyridine, 97%, Thermo Scientific Chemicals
Catalog numberH66177.03
Price (USD)/ Each
39.20
-
Add to cart
Chemical Identifiers
CAS139585-70-9
IUPAC Name6-bromopyridine-3-carbonitrile
Molecular FormulaC6H3BrN2
InChI KeyXHYGUDGTUJPSNX-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Assay (unspecified)>96.0%
FormCrystals or crystalline powder
Water Content (Karl Fischer Titration)<1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only