Thermo Scientific Chemicals

4-Aminoimidazole-5-carboxamide, 95%, Thermo Scientific Chemicals

Catalog number: H66184.06
5 g, Each
Thermo Scientific Chemicals

4-Aminoimidazole-5-carboxamide, 95%, Thermo Scientific Chemicals

Catalog number: H66184.06
5 g, Each
Quantity
Catalog number: H66184.06
also known as H66184-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
360-97-4
IUPAC Name
4-amino-1H-imidazole-5-carboxamide
Molecular Formula
C4H6N4O
InChI Key
DVNYTAVYBRSTGK-UHFFFAOYSA-N
SMILES
NC(=O)C1=C(N)N=CN1
Assay (HPLC)
>94.0%

Description

4-Aminoimidazole-5-carboxamide is used as Catalytic agent, Petrochemical additive. It is metabolite of temozolomide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminoimidazole-5-carboxamide is used as Catalytic agent, Petrochemical additive. It is metabolite of temozolomide.

Solubility
Soluble in DMSO, Methanol, Water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text