4-Aminoimidazole-5-carboxamide, 95%, Thermo Scientific Chemicals
4-Aminoimidazole-5-carboxamide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminoimidazole-5-carboxamide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H66184.14
also known as H66184-14
Price (USD)/ Each
164.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
164.00
Add to cart
4-Aminoimidazole-5-carboxamide, 95%, Thermo Scientific Chemicals
Catalog numberH66184.14
Price (USD)/ Each
164.00
-
Add to cart
Chemical Identifiers
CAS360-97-4
IUPAC Name4-amino-1H-imidazole-5-carboxamide
Molecular FormulaC4H6N4O
InChI KeyDVNYTAVYBRSTGK-UHFFFAOYSA-N
SMILESNC(=O)C1=C(N)N=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
4-Aminoimidazole-5-carboxamide is used as Catalytic agent, Petrochemical additive. It is metabolite of temozolomide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminoimidazole-5-carboxamide is used as Catalytic agent, Petrochemical additive. It is metabolite of temozolomide.

Solubility
Soluble in DMSO, Methanol, Water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

General References:

  1. Glenn E. Housholder; Ti Li Loo. Elevated urinary excretion of 4-aminoimidazole-5-carboxamide in patients after intravenous injection of 4-(3, 3-dimethyl-1-triazeno) imidazole-5-carboxamide. Life Sciences.1969, 8, 533-536.
  2. Jeffrey D. Newman; Bruce W. Schultz and K. Dale Noel. Dissection of Nodule Development by Supplementation of Rhizobium leguminosarum biovar phaseoli Purine Auxotrophs with 4-Aminoimidazole-5-Carboxamide Riboside. Plant Physiology.1992, 99, (2), 401-408.