7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%, Thermo Scientific Chemicals
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H66188.06
also known as H66188-06
Price (USD)/ Each
222.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
262.00 
Save 39.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
222.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
262.00 
Save 39.35 (15%)
Add to cart
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 95%, Thermo Scientific Chemicals
Catalog numberH66188.06
Price (USD)/ Each
222.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
262.00 
Save 39.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1810-74-8
IUPAC Name7-methoxy-2,2,4-trimethyl-1,2-dihydroquinoline
Molecular FormulaC13H17NO
InChI KeyVNIQAUZZZWOJPT-UHFFFAOYSA-N
SMILESCOC1=CC2=C(C=C1)C(C)=CC(C)(C)N2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.00%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only