Thermo Scientific Chemicals

N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H66193.03
1 g, Each
Thermo Scientific Chemicals

N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H66193.03
1 g, Each
Quantity
Catalog number: H66193.03
also known as H66193-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
286961-14-6
IUPAC Name
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate
Molecular Formula
C16H28BNO4
InChI Key
VVDCRJGWILREQH-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1
Assay (HPLC)
>94.0%

Description

Reagent used for suzuki-Miyaura cross-coupling using palladium phosphine catalyst, palladium-catalyzed ligand-controlled regioselective Suzuki coupling, wrenchnolol derivative optimized for gene activation in cells. Also used in Preparation of several enzymatic inhibitors and receptor ligands, orally active anaplastic lymphoma kinase inhibitors and oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for suzuki-Miyaura cross-coupling using palladium phosphine catalyst, palladium-catalyzed ligand-controlled regioselective Suzuki coupling, wrenchnolol derivative optimized for gene activation in cells. Also used in Preparation of several enzymatic inhibitors and receptor ligands, orally active anaplastic lymphoma kinase inhibitors and oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes.

Solubility
Soluble in most organic solvents. Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text