2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-1-cyclopentene-1-carbonitrile s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Incompatible with heat and oxidizing agents.
RUO – Research Use Only
J. P. Ferris; R. W. Trimmer. Chemical evolution. XXVIII. Photochemical conversion of enaminonitriles to imidazoles. Scope and mechanism. J. Org. Chem.1976, 41, (1), 19-24.