3-Bromo-1-(phenylsulfonyl)indole, 97%, Thermo Scientific Chemicals
3-Bromo-1-(phenylsulfonyl)indole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-1-(phenylsulfonyl)indole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H66226.06
also known as H66226-06
Price (USD)/ Each
91.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
108.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
91.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
108.00 
Save 16.35 (15%)
Add to cart
3-Bromo-1-(phenylsulfonyl)indole, 97%, Thermo Scientific Chemicals
Catalog numberH66226.06
Price (USD)/ Each
91.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
108.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS99655-68-2
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Assay (HPLC)≥96.0%
Melting Point117-119?C
Appearance (Color)Brown
Proton NMR≥96.0%. Conforms to structure,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only