6-Methoxyindole-2-carboxylic acid, 95%, Thermo Scientific Chemicals
6-Methoxyindole-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methoxyindole-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66239.03
also known as H66239-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS16732-73-3
IUPAC Name6-methoxy-1H-indole-2-carboxylic acid
Molecular FormulaC10H9NO3
InChI KeyXNBGANWAZJWOHS-UHFFFAOYSA-N
SMILESCOC1=CC=C2C=C(NC2=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light gray
FormPowder
Assay from Supplier's CofA≥94.0%
Proton NMRConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only