1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%, Thermo Scientific Chemicals
1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H66242.06
also known as H66242-06
Price (USD)/ Each
270.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
270.00
Add to cart
1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH66242.06
Price (USD)/ Each
270.00
-
Add to cart
Specifications
Chemical Name or Material1-(Cyclopropylcarbonyl)piperazine hydrochloride
CAS1021298-67-8
Melting Point175°C to 179°C
ColorWhite
Molecular FormulaC8H15ClN2O
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only