4-Bromo-3-hydroxybenzyl alcohol, 95%, Thermo Scientific Chemicals
4-Bromo-3-hydroxybenzyl alcohol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-hydroxybenzyl alcohol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66253.MD
also known as H66253-MD
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS2737-19-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.60%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only