Thermo Scientific Chemicals

N-Isopropylacrylamide, 97%, Thermo Scientific Chemicals

Catalog number: H66262.14
25 g, Each
Thermo Scientific Chemicals

N-Isopropylacrylamide, 97%, Thermo Scientific Chemicals

Catalog number: H66262.14
25 g, Each
Quantity
Catalog number: H66262.14
also known as H66262-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2210-25-5
IUPAC Name
N-(propan-2-yl)prop-2-enamide
Molecular Formula
C6H11NO
InChI Key
QNILTEGFHQSKFF-UHFFFAOYSA-N
SMILES
CC(C)NC(=O)C=C
Form
Solid
Appearance (Color)
White
Assay (HPLC)
>96.0%

Description

N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text