5-(Fmoc-amino)valeric acid, 98%, Thermo Scientific Chemicals
5-(Fmoc-amino)valeric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-(Fmoc-amino)valeric acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H66264.06
also known as H66264-06
Price (USD)/ Each
148.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
148.00
Add to cart
5-(Fmoc-amino)valeric acid, 98%, Thermo Scientific Chemicals
Catalog numberH66264.06
Price (USD)/ Each
148.00
-
Add to cart
Chemical Identifiers
CAS123622-48-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
5-(Fmoc-amino)valeric acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Fmoc-amino)valeric acid is used as pharmaceutical intermediate.

Notes
Protect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with
RUO – Research Use Only