Chemical Identifiers
CAS188715-40-4
IUPAC Name4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]cyclohexane-1-carboxylic acid
Molecular FormulaC23H25NO4
InChI KeyMLMIBGARTUSGND-UHFFFAOYSA-N
SMILESOC(=O)C1CCC(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)CC1
View more